Order Entry
Poland
ContactUsLinkComponent
 
 
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
 
undefined
 
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • Loss on drying (300°C):
    Max. 1 %
  • Water insoluble matter:
    Max. 0.01 %
  • Silver (Ag):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 5 ppm
  • Arsenic (As):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 200 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 5 ppm
  • Thallium (Tl):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 40 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Silicate (as SiO₂):
    Max. 50 ppm
  • Chloride (Cl):
    Max. 30 ppm
  • Phosphate (PO₄):
    Max. 10 ppm
  • Total nitrogen (as N):
    Max. 10 ppm

 

 

Frequently Bought Together