Cyclopentane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Pentamethylene
CAS Number:287-92-3
:
Formula:C₅H₁₀
MDL Number:MFCD00001356
Boiling Pt:47…49 °C
Melting Pt:–95 °C
Density:0.748
Molecular Weight:70.13 g/mol
Flash Pt:–37 °C (–34 °F)
UN:1146
ADR:3 II
Merck Index:13,02769
EINECS:206-016-6
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together