Scandium(III) trifluoromethanesulphonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:144026-79-9
:
Formula:C₃F₉O₉S₃Sc
MDL Number:MFCD00192433
:492.17 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together