Bis(triphenylphosphine)palladium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13965-03-2
:
Formula:C₃₆H₃₀Cl₂P₂Pd
MDL Number:MFCD00009593
:701.91 g/mol
:M-Clarity™ quality level = MQ100
EINECS:237-744-2
Frequently Bought Together