Order Entry
Poland
ContactUsLinkComponent
Methyl-2,5-dichlorobenzoate, Sigma-Aldrich®
Methyl-2,5-dichlorobenzoate, Sigma-Aldrich®
Catalog # SIAL166731-10G
Supplier:  Merck
CAS Number:  
Methyl-2,5-dichlorobenzoate, Sigma-Aldrich®
Catalog # SIAL166731-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :205.04 g/mol
EINECS:220-815-7
CAS Number:2905-69-3
Formula:C₈H₆Cl₂O₂
MDL Number:MFCD00000606