Order Entry
Poland
ContactUsLinkComponent
4-Chloropyridine hydrochloride
4-Chloropyridine hydrochloride
Catalog # MOLEM89759763
Supplier:  Molekula
CAS Number:  
4-Chloropyridine hydrochloride
Catalog # MOLEM89759763
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :150.01 g/mol
EINECS:230-946-1
CAS Number:7379-35-3
Formula:C₅H₅Cl₂N
MDL Number:MFCD00012829