D-(-)-Quinic acid, Sigma-Aldrich®
8.00218.0050
: Merck
:
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
- Solubility:290 g/l (9 °C) (experimental)
- Melting Point:162.5 °C
- Molar Mass:192.17 g/mol
- Bulk Density:670 - 750 kg/m3
- Density:1.64 g/cm3 (20 °C)
- pH:2.0 (10 g/l, H2O, 20 °C)
:
:192.17 g/mol
:M-Clarity™ quality level = MQ200
:201-072-8
:77-95-2
:C₇H₁₂O₆
:MFCD00003864