

4-Picolyl chloride hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1822-51-1
:
Formula:C₆H₇Cl₂N
MDL Number:MFCD00012826
Melting Pt:160…163 °C
Storage Temperature:Ambient
EINECS:217-350-7
UN:3261
ADR:8,III
:164.03 g/mol
:GHS07
:GHS05
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥96.0 %
- GC:≥99.4 %
- Color scale:≤20 APHA
Specification Test Results
Frequently Bought Together