Order Entry
Poland
ContactUsLinkComponent
3-Picolyl chloride hydrochloride 99%
Corrosive
Irritant
3-Picolyl chloride hydrochloride 99%
Certificates
undefined
Corrosive
Irritant
3-Picolyl chloride hydrochloride 99%
Specification Test Results
  • Appearance:
    White to beige Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥98.5 %
  • GC:
    ≥99.4 %
  • Color scale:
    ≤20 APHA