Order Entry
Poland
Orders LinkContactUsLinkComponent
Quinaldic acid

Quinaldic acid

25 g
Catalog # MOLE15641920-25G
Irritant
Marketsource
Supplier:   Molekula
Quinaldic acid
Catalog # MOLE15641920-25G
Supplier:  Molekula
Marketsource
CAS Number:  93-10-7
Quinaldic acid

25 g

CAS Number:  93-10-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
  • CAS Number:93-10-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00006752
  • Molecular Weight:173.17 g/mol
  • Merck Index:12,08227
  • Melting Pt:154…157 °C
  • Storage Temperature:Ambient
  • EINECS:202-218-3
Specifications

Specifications

  • Pk:
    25 g

Specifications