Quinaldic acid ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 93-10-7 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 156-159° Beilstein Reference: 126322 Merck Reference: 14,8046 MDL No.: MFCD00006752
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Merck Index:14,8046
- Melting Pt:156…159 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 93-10-7 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Melting Point: 156-159° Beilstein Reference: 126322 Merck Reference: 14,8046 MDL No.: MFCD00006752
Synonyms: Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
- CAS Number:93-10-7
- Formula:C₁₀H₇NO₂
- MDL Number:MFCD00006752
- Molecular Weight:173.17 g/mol
- Merck Index:14,8046
- Melting Pt:156…159 °C
- Storage Temperature:Ambient
- EINECS:202-218-3
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



