Stirring blades, moon-shaped
Supplier: Bohlender
Certificates
About this item
Stirrer blades made of PTFE, with double sided groove.
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Suitable for glass stirrer shafts with double pivot, KPG glass stirrer shafts and PTFE-jacketed stainless steel stirrer shafts.
Frequently Bought Together