Order Entry
Poland
ContactUsLinkComponent
Phosgene 20% in toluene, Sigma-Aldrich®
Phosgene 20% in toluene, Sigma-Aldrich®
Catalog # SIAL79380-500ML
Supplier:  Merck
CAS Number:  
Phosgene 20% in toluene, Sigma-Aldrich®
Catalog # SIAL79380-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :98.92
EINECS:200-870-3
UN:3384
ADR:6.1,I
CAS Number:75-44-5
Formula:CCl₂O