
Triphenylene-[D₁₂], Sigma-Aldrich®
Catalog # SIAL616362-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.2 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS05
CAS Number:17777-56-9
Formula:C₁₈D₁₂