Benzoin methyl ether, Sigma-Aldrich®
Catalog # SIAL479314-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:226.27 g/mol
EINECS:222-538-7
CAS Number:3524-62-7
Formula:C₁₅H₁₄O₂
MDL Number:MFCD00008492