Order Entry
Poland
ContactUsLinkComponent
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers), Sigma-Aldrich®
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL375438-25G
Supplier:  Merck
CAS Number:  
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers), Sigma-Aldrich®
Catalog # SIAL375438-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :172.18 g/mol
EINECS:432-490-0
CAS Number:3971-31-1
Formula:C₈H₁₂O₄
MDL Number:MFCD00134411