Order Entry
Poland
ContactUsLinkComponent
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Catalog # MOLE50547934-100G
Supplier:  Molekula
CAS Number:  
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Catalog # MOLE50547934-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :462.08
EINECS:206-192-4
CAS Number:306-94-5
Formula:C₁₀F₁₈
MDL Number:MFCD00010626