3-(Dimethylamino)propiophenone hydrochloride, Sigma-Aldrich®
Catalog # 8.41828.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Melting Point:155 - 156 °C
- Molar Mass:213.7 g/mol
- Hill Formula:C11H16ClNO
Specification Test Results
About this item
:
:213.71 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:212-909-1
UN:2811
CAS Number:879-72-1
Formula:C₁₁H₁₆ClNO
MDL Number:MFCD00012481