Triphenylene-[D₁₂] (98% D)
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
CAS Number:17777-56-9
:
Formula:C₁₈D₁₂
:240.2 g/mol
UN:3077
ADR:9,III
:GHS05
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Folia uszczelniająca do aplikacji ręcznej, Parafilm® M
Supplier: HEATHROW
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Woda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Rękawice nitrylowe, NITRILE
Supplier: VWR Collection
VWR® Chusteczki do twarzy
Supplier: VWR Collection
Pask wskaśnikowe pH, MQuant®, Supelco®
Supplier: Merck
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Rękawice nitrylowe
Supplier: VWR Collection
VWR®, Pipety Pasteura, High Performance
Supplier: VWR Collection
Rękawice nitrylowe, Kimtech™ Purple Nitrile™, Kimberly-Clark
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection