Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
α,2-Dichlorotoluene 98+%
 
undefined
 
 
 
α,2-Dichlorotoluene 98+%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5580 to 1.5600 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Residue after ignition:
    ≤0.1 %

 

 

Frequently Bought Together