Butyrophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:495-40-9
:
Formula:C₆H₅C(O)(CH₂)₂CH₃
MDL Number:MFCD00009397
:148.2 g/mol
Boiling Pt:222 °C
Melting Pt:11…13 °C
Density:1.021
Flash Pt:88 °C (190 °F)
Storage Temperature:Ambient
EINECS:207-799-7
Related Information
Frequently Bought Together