Order Entry
Poland
ContactUsLinkComponent
4-Cyanophenylboronic acid pinacol ester 97%
Irritant
4-Cyanophenylboronic acid pinacol ester 97%
Certificates
undefined
Irritant
4-Cyanophenylboronic acid pinacol ester 97%

Specification Test Results

  • Appearance:
    Light yellow to orange to tan Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    94 to 99 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together