



Trimethylsilyl isocyanate ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1118-02-1
:
Formula:C₄H₉NOSi
MDL Number:MFCD00001993
:115.21 g/mol
Boiling Pt:90…92 °C
Melting Pt:–49 °C
Density:0.859
Flash Pt:–2 °C (28 °F)
EINECS:214-256-8
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together