


Trimethylsilyl isocyanate 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1118-02-1
:
Formula:C₄H₉NOSi
MDL Number:MFCD00001993
:115.21 g/mol
Melting Pt:–49 °C
Flash Pt:–2 °C
EINECS:214-256-8
UN:2478
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3970 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together