


2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4274-38-8
:
Formula:C₇H₇ClF₃NS
MDL Number:MFCD00042600
:229.65
Melting Pt:186…190 °C
Density:2.46
EINECS:429-560-8
ADR:8,II
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together