Order Entry
Poland
ContactUsLinkComponent
 
5-Methoxytryptamine hydrochloride
Catalog # MOLEM45320712
Supplier:  Molekula
CAS Number:  
5-Methoxytryptamine hydrochloride
Catalog # MOLEM45320712
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.71 g/mol
EINECS:200-637-6
CAS Number:66-83-1
Formula:C₁₁H₁₅ClN₂O
MDL Number:MFCD00005662