Order Entry
Poland
ContactUsLinkComponent
 
Tetrachloromethane
Catalog # PROOCHE04
CAS Number:  
undefined
Tetrachloromethane
Catalog # PROOCHE04
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :153.82 g/mol
Merck Index:12,01864
Boiling Pt:76.7 °C (1013 hPa)
Melting Pt:–23 °C
Density:1.5942 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-262-8
UN:1846
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:56-23-5
Formula:CCl₄