Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(2-Fluorophenyl)-1H-tetrazole
5-(2-Fluorophenyl)-1H-tetrazole
5-(2-Fluorophenyl)-1H-tetrazole
5-(2-Fluorophenyl)-1H-tetrazole
MOLE15844514-1G
:
Molekula
:
5-(2-Fluorophenyl)-1H-tetrazole
MOLE15844514-1G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
164.14 g/mol
:
50907-19-2
:
MFCD00665794
:
C₇H₅FN₄
:
MFCD00798373