
Methylene blue for microscopical staining
Supplier: CHEMPUR
Certificates
About this item
CAS Number:61-73-4
:MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008
:319.86 g/mol
Merck Index:12,06137
Melting Pt:180 °C
Density:1.757 g/cm³ (22 °C)
EINECS:200-515-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together