Order Entry
Poland
ContactUsLinkComponent
2-Methoxyethyl isothiocyanate, Sigma-Aldrich®
2-Methoxyethyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL709123-5G
Supplier:  Merck
CAS Number:  
2-Methoxyethyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL709123-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:88.5…92 °C (23.5 mmHg)
Density:1,08 g/cm³ (20 °C)
 :117.17 g/mol
EINECS:254-070-4
CAS Number:38663-85-3
Formula:C₄H₇NOS
MDL Number:MFCD00041126
Boiling Pt:88,5…92 °C (23.5 mmHg)