Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 25%, Technical Grade
 :   CHEMPUR
Ammonia solution 25%, Technical Grade
 :  CHEMPUR
Ammonia solution 25%, Technical Grade
 :  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  •  :1336-21-6
  •  :NH₃
  •  :MFCD00066650
  •  :35.05 g/mol
  •  :32…37.7 °C (1013 hPa)
  •  :–60…–57.5 °C
  •  :0.88…0.91 g/cm³ (20 °C)
  •  :Ambient
  •  :215-647-6
  •  :2672
  •  :8,III
Product Family Options