Order Entry
Poland
ContactUsLinkComponent
2,6-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Irritant
2,6-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIALA89502-5G
Supplier:  Merck
CAS Number:  
Irritant
2,6-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIALA89502-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :240.22 g/mol
Melting Pt:> 330 °C
 :M-Clarity™ quality level = MQ100
EINECS:201-544-3
 :GHS07
CAS Number:84-60-6
Formula:C₁₄H₈O₄
MDL Number:MFCD00001228