Order Entry
Poland
ContactUsLinkComponent
12-Bromo-1-dodecanol, Sigma-Aldrich®
12-Bromo-1-dodecanol, Sigma-Aldrich®
Catalog # SIAL224677-5G
Supplier:  Merck
CAS Number:  
12-Bromo-1-dodecanol, Sigma-Aldrich®
Catalog # SIAL224677-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :265.23 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:3344-77-2
Formula:HO(CH₂)₁₁CH₂Br
MDL Number:MFCD00004754