

1-Iodopropane stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-08-4
:
Formula:C₃H₇I
MDL Number:MFCD00001095
:169.99 g/mol
Merck Index:12,08047
Boiling Pt:103 °C (1013 hPa)
Melting Pt:–98 °C
Density:1.741 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:44 °C
:Stabilised with sodium carbonate (Na₂CO₃)
Storage Temperature:Ambient
EINECS:203-460-2
UN:2392
:GHS02
:GHS06
Specification Test Results
- Solubility:1.1 g/l (20 °C)
- Melting Point:-98 °C
- Molar Mass:169.99 g/mol
- Boiling Point:103 °C (1013 mbar)
- Vapor Pressure:57 hPa (25 °C)
- Flash Point:44 °C
- Refractive Index:1.5055 (20 °C, 589 nm)
- Density:1.75 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together