Order Entry
Poland
Orders LinkContactUsLinkComponent
Guaiacol

Guaiacol

Glass bottle for solids, 1 kg
Catalog # MOLEM38189717
Irritant
Marketsource
Supplier:   Molekula
Guaiacol
Catalog # MOLEM38189717
Supplier:  Molekula
Marketsource
CAS Number:  90-05-1
Guaiacol

Glass bottle for solids, 1 kg

CAS Number:  90-05-1
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
  • CAS Number:90-05-1
  • Formula:C₇H₈O₂
  • MDL Number:MFCD00002185
  • Molecular Weight:124.14 g/mol
  • Merck Index:13,04568
  • Boiling Pt:205 °C (1013 hPa)
  • Melting Pt:28…32 °C
  • Density:1.1534 g/cm³ (0 °C)
  • Flash Pt:82 °C
  • Storage Temperature:Ambient
  • EINECS:201-964-7
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.