


1,3-Dinitrobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:99-65-0
:
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007222
:168.11 g/mol
Boiling Pt:276 °C (1013 hPa)
Melting Pt:90 °C
Density:1.55 g/cm³ (20 °C)
Flash Pt:150 °C
Storage Temperature:Ambient
UN:3443
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:202-776-8
:GHS06
:GHS08
:GHS09
Frequently Bought Together