Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethylaniline 99%
Toxic
Health
Environment
N,N-Dimethylaniline 99%
Certificates
undefined
Toxic
Health
Environment
N,N-Dimethylaniline 99%
Certificates

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5570 to 1.5590 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together