1,4-Difluorobenzene
Catalog # MOLEM43548945
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:114.09 g/mol
Merck Index:13,03172
Boiling Pt:88 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-742-9
UN:1993
ADR:3,II
CAS Number:540-36-3
Formula:C₆H₄F₂
MDL Number:MFCD00000344