Order Entry
Poland
Orders LinkContactUsLinkComponent
Accessories for Cooling Box

Accessories for Cooling Box

HDPE cooling block for Kartell Frigo box, 500 ml, 17,5×9×3 cm
Catalog # KART3707
Supplier:   KARTELL
Accessories for Cooling Box
Catalog # KART3707
Supplier:  KARTELL
Accessories for Cooling Box
Accessories for Cooling Box

HDPE cooling block for Kartell Frigo box, 500 ml, 17,5×9×3 cm

Price
30,40 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
Specifications
  • Capacity:
    500 ml
  • Description:
    HDPE cooling block for Kartell Frigo box
  • W×D×H:
    17,5×9×3 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.