1,2,3,4-Tetrafluorobenzene
Catalog # MOLEM21602144
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:150.08 g/mol
Boiling Pt:95 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.811 g/cm³ (20 °C)
EINECS:208-997-6
UN:1993
ADR:3,II
CAS Number:551-62-2
Formula:C₆H₂F₄
MDL Number:MFCD00000285