

1,4-Dichlorobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-46-7
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000604
:147 g/mol
Merck Index:13,03082
Boiling Pt:173.4 °C (1013 hPa)
Melting Pt:48…50 °C
Density:1.458 g/cm³ (20 °C)
EINECS:203-400-5
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
Flash Pt:65 °C
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together