Salicylic acid ≥98%, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
- CAS Number:69-72-7
- Formula:(HO)C₆H₄CO₂H
- MDL Number:MFCD00002439
- Molecular Weight:138.12 g/mol
- Merck Index:13,08411
- Boiling Pt:336 °C (1013 hPa)
- Melting Pt:157…159 °C
- Density:1.443 g/cm³ (20 °C)
- Flash Pt:157 °C
- Storage Temperature:Ambient
- EINECS:200-712-3
Documents
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
P20_VWRC20659.295_SPEC_EN.pdf spec_VWRC20659.295 spec_VWRC20659.295
Product specification
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



