Order Entry
Poland
ContactUsLinkComponent
 
 
 
 
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
    
 
undefined
 
 
 
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (calculated on anhydrous):
    Min. 99.9 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.05 %

 

Frequently Bought Together