The Wheaton® R2P™ 2.0 roller culture apparatus is designed to allow for the development and growth of cultured cells from research to production quantities without having to change equipment and revalidate protocols. The R2P™ 2.0 system utilises computerised measurement of the actual bottle speed and allows networking for up to 255 units. Applications include, Viruses for vaccines or related studies, proteins, enzymes, antibodies, live cell assays.
- Robust touch screen interface that is easy to view from a distance
- Readily recognised icons for intuitive navigation
- Digitally controlled maintenance-free motor accurate to 0,01 min⁻¹
- Capable of remote interface and monitoring through SCADA systems
- Multiple decking and capacity configurations available for process flexibility
Two deck configurations are available. Production spacing with 152 mm between decks maximises use of incubator space, whilst modular spacing with 181 mm between decks allows easier removal and replacement of bottles, reducing disruption to cells. Systems with five placement areas per deck allow use with an upright incubator, whilst eight place systems are designed for large scale production installations.
- Catalog No:
- WHEAWSBMR5010-D
- WHEAWSBMR5020-D
- WHEAWRBMR5031-C
- WHEAWSBMR5011-D
- WHEAWRBMR5071-C
- 392-0354
- WHEAWSTPF8111-C
- WHEAWSTPF8110-C
- 392-0351
- WHEAWRBPR5030-B
- WHEAWRBPR5030-A
- WHEAWRBPF5110-J
- WHEAWRBPR5030-F
- 392-0352
- WHEAWSBMR5010-C
- WHEAWSTPF5111-C
- 392-0353
- WHEAWRTPF8110-F
- WHEAWRTPF8110-J
- WHEAWRBMR5030-J
- WHEAWRTPF5110-A
- WHEAWRTPF5110-B
- WHEAWRBPR5030-J
- WHEAWRTPF8110-B
- WHEAWRTPF8110-A
- WHEAWRBMR5010-F
- WHEAWRBPR5050-C
- 392-0356
- WHEAWRBMR5010-D
- 392-0355
- WHEAWRBMR5010-C
- WHEAWRTPF5110-J
- WHEAWRBMR5010-J
- WHEAWRBMR5091-C
- 392-0349
- WHEAWRBPF8110-C
- WHEAWSBMR5040-D
- 392-0454
- WHEAWRBMR5030-B
- WHEAWRBMR5010-B
- WHEAWRBMR5030-A
- WHEAWSBPF5110-C
- WHEAWSBMR5040-C
- WHEAWRBPF5110-F
- 392-0453
- 392-0452
- 392-0455
- WHEAWRBMR5090-B
- WHEAWRBMR5090-A
- 392-0350
- WHEAWRBPF8111-C
- WHEAWRBPF5111-C
- WHEAWRBMR5090-F
- 392-0456
- WHEAWRBMR5030-F
- WHEAWRBMR5010-A
- WHEAWRBPF5110-A
- WHEAWRBMR5090-J
- WHEAWRTMR5071-C
- WHEAWRTPF5110-F
- WHEAWRBPF5110-B
- 392-0458
- Model:
- WSBMR5010
- WSBMR5020
- WRBMR5031
- WSBMR5011
- WRBMR5071
- WRTPF5110
- WSTPF8111
- WSTPF8110
- WRBPR5030
- WRBPR5030
- WRBPR5030
- WRBPF5110
- WRBPF5030
- WRBPR5030
- WSBMR5010
- WSTPF5111
- WRTPF5110
- WRTPF8110
- WRTPF8110
- WRBMR5030
- WRTPF5110
- WRTPF5110
- WRBPF5030
- WRTPF8110
- WRTPF8110
- WRBMR5010
- WRBPR5050
- WRTPF8110
- WRBMR5010
- WRTPF8110
- WRBMR5010
- WRTPF5110
- WRBMR5010
- WRBMR5091
- WRBPF5110
- WRBPF8110
- WSBMR5040
- WRBMR5070
- WRBMR5030
- WRBMR5010
- WRBMR5030
- WSBPF5110
- WSBMR5040
- WRBPF5110
- WRBMR5030
- WRBMR5030
- WRBMR5090
- WRBMR5090
- WRBMR5090
- WRBPF5110
- WRBPF8111
- WRBPF5111
- WRBMR5090
- WRBMR5090
- WRBMR5030
- WRBMR5010
- WRBPF5110
- WRBMR5090
- WRTMR5071
- WRTPF5110
- WRBPF5110
- WSBMR5090
- Type:
- Bottom drive modular standard spacing, removable deck
- Bottom drive modular standard spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular standard spacing, removable deck
- Bottom drive modular spacing, removable deck
- Top drive production spacing, fixed deck
- Top drive production spacing
- Top drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive modular standard spacing, removable deck
- Top drive production spacing, fixed deck
- Top drive production spacing, fixed deck
- Top drive production spacing
- Top drive production spacing
- Bottom drive modular spacing, removable deck
- Top drive production spacing, fixed deck
- Top drive production spacing, fixed deck
- Bottom drive production spacing
- Top drive production spacing, fixed deck
- Top drive production spacing, fixed deck
- Bottom drive modular spacing, removable deck
- Bottom drive production spacing
- Top drive production spacing, fixed deck
- Bottom drive modular spacing, removable deck
- Top drive production spacing, fixed deck
- Bottom drive modular spacing, removable deck
- Top drive production spacing, fixed deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive modular standard spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive production spacing
- Bottom drive modular standard spacing, removable deck
- Bottom drive production spacing
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive production spacing
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive modular spacing, removable deck
- Bottom drive production spacing
- Bottom drive modular spacing, removable deck
- Top drive production spacing
- Top drive production spacing, fixed deck
- Bottom drive production spacing
- Bottom drive modular standard spacing, removable deck
- Bottle capacity:
- 5 bottle positions
- 10 bottle positions
- 15 bottle positions
- 5 bottle positions
- 35 bottle position
- 52 bottle positions
- 86 bottle positions
- 86 bottle positions
- 15 bottle positions
- 15 bottle positions
- 15 bottle positions
- 55 bottle positions
- 15 bottle positions
- 15 bottle positions
- 5 bottle positions
- 52 bottle positions
- 52 bottle positions
- 85 bottle positions
- 85 bottle positions
- 15 bottle position
- 52 bottle positions
- 52 bottle positions
- 15 bottle positions
- 85 bottle positions
- 85 bottle positions
- 5 bottle positions
- 25 bottle positions
- 85 bottle positions
- 5 bottle positions
- 85 bottle positions
- 5 bottle positions
- 52 bottle positions
- 5 bottle positions
- 45 bottle position
- 55 bottle positions
- 88 bottle positions
- 20 bottle positions
- 35 bottle positions
- 15 bottle position
- 5 bottle positions
- 5 bottle positions
- 55 bottle positions
- 20 bottle positions
- 55 bottle positions
- 15 bottle position
- 15 bottle position
- 45 bottle position
- 45 bottle position
- 45 bottle positions
- 55 bottle positions
- 88 bottle positions
- 55 bottle positions
- 45 bottle position
- 45 bottle position
- 15 bottle position
- 5 bottle positions
- 55 bottle positions
- 45 bottle position
- 35 bottle positions
- 52 bottle positions
- 55 bottle positions
- 45 bottle positions
- Accuracy:
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- 0,01 min⁻¹
- Drive:
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Belt driven
- Motor:
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Rotation:
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking; rotation alarm included
- Direction: Clockwise, counter-clockwise and rocking
- Interfaces:
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Modbus® and Wheaton protocols via ethernet and RS422
- Electrical:
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 120 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- 240 VAC, 50/60 Hz, 35 W
- 100 VAC, 50/60 Hz, 35 W
- 230 VAC, 50/60 Hz, 35 W
- Max. humidity:
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- Operating temperature:
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- 10...40 °C
- W×D×H:
- 760×630×340 mm
- 760×630×340 mm
- 760×630×740 mm
- 760×630×340 mm
- 760×630×1450 mm
- 790×630×1900 mm
- 1200×630×1900 mm
- 1200×630×1900 mm
- 790×630×650 mm
- 790×630×650 mm
- 790×630×650 mm
- 790×630×1870 mm
- 790×630×650 mm
- 790×630×650 mm
- 760×630×340 mm
- 790×630×1900 mm
- 790×630×1900 mm
- 1200×630×1900 mm
- 1200×630×1900 mm
- 760×630×510 mm
- 790×630×1900 mm
- 790×630×1900 mm
- 790×630×650 mm
- 1200×630×1900 mm
- 1200×630×1900 mm
- 760×630×340 mm
- −
- 1200×630×1900 mm
- 760×630×340 mm
- 1200×630×1900 mm
- 760×630×340 mm
- 790×630×1900 mm
- 760×630×340 mm
- 760×630×1080 mm
- 790×630×1870 mm
- 1200×630×1870 mm
- 760×630×920 mm
- 760×630×1450 mm
- 760×630×510 mm
- 760×630×340 mm
- 760×630×510 mm
- 790×630×1870 mm
- 760×630×920 mm
- 790×630×1870 mm
- 760×630×510 mm
- 760×630×510 mm
- 760×630×1080 mm
- 760×630×1080 mm
- 760×630×1080 mm
- 790×630×1870 mm
- 1200×630×1870 mm
- 790×630×1870 mm
- 760×630×1080 mm
- 760×630×1080 mm
- 760×630×510 mm
- 760×630×340 mm
- 790×630×1870 mm
- 760×630×1080 mm
- 790×630×1450 mm
- 790×630×1900 mm
- 790×630×1870 mm
- 790×630×1790 mm
- Weight:
- 32 kg
- 32 kg
- 51 kg
- 32 kg
- 89 kg
- 113 kg
- 159 kg
- 159 kg
- 39 kg
- 39 kg
- 39 kg
- 99 kg
- 39 kg
- 39 kg
- 32 kg
- 113 kg
- 113 kg
- 159 kg
- 159 kg
- 51 kg
- 113 kg
- 113 kg
- 39 kg
- 159 kg
- 159 kg
- 32 kg
- −
- 159 kg
- 32 kg
- 159 kg
- 32 kg
- 113 kg
- 32 kg
- 108 kg
- 99 kg
- 167 kg
- −
- 89 kg
- 51 kg
- 32 kg
- 51 kg
- 99 kg
- −
- 99 kg
- 51 kg
- 51 kg
- 108 kg
- 108 kg
- 108 kg
- 99 kg
- 167 kg
- 99 kg
- 108 kg
- 108 kg
- 51 kg
- 32 kg
- 99 kg
- 108 kg
- 89
- 113 kg
- 99 kg
- 108 kg