Order Entry
Poland
ContactUsLinkComponent
Benzonitrile for synthesis, Sigma-Aldrich®
Irritant
Benzonitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Benzonitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    -13 °C
  • Molar Mass:
    103.12 g/mol
  • Boiling Point:
    190 °C (1013 hPa)
  • Vapor Pressure:
    0.7 hPa (20 °C)
  • Flash Point:
    70 °C
  • Refractive Index:
    1.5289 (20 °C, 589 nm)
  • Explosion Limit:
    1.4 - 7.2 %(V)
  • Density:
    1.00 g/cm3 (20 °C)
  • Ignition Point:
    550 °C (experimental)

Specification Test Results

Related Information

Frequently Bought Together