Order Entry
Poland
ContactUsLinkComponent
Water purification systems, RiOs™ 30/50/100/150/200
 :  Merck
Water purification systems, RiOs™ 30/50/100/150/200
 :  Merck
  • Catalog No:
  • MILLZR0S50050SAVT
  • MILLZR0S50200SAVT
  • MILLZR0S50150SAVT
  • MILLZR0S50100SAVT
  • MILLZR0S50030SAVT
  • Model:
  • RiOs 50
  • RiOs 200
  • RiOs 150
  • RiOs 100
  • RiOs 30
  • Feed water:
  • Tap water (potable)
  • Tap water (potable)
  • Tap water (potable)
  • Tap water (potable)
  • Tap water (potable)
  • Production flow rate:
  • 50 L/h
  • 200 L/h
  • 150 L/h
  • 100 L/h
  • 30 L/h
  • TOC content:
  • < 50 ppb
  • < 50 ppb
  • < 50 ppb
  • < 50 ppb
  • < 50 ppb
  • Bacteria:
  • < 10 CFU/ml
  • < 10 CFU/ml
  • < 10 CFU/ml
  • < 10 CFU/ml
  • < 10 CFU/ml
  • Particles:
  • > 99 %
  • > 99 %
  • > 99 %
  • > 99 %
  • > 99 %
  • W×D×H:
  • 662×441×734 mm
  • 662×441×734 mm
  • 662×441×734 mm
  • 662×441×734 mm
  • 662×441×734 mm
  • Weight:
  • 34 kg
  • 40 kg
  • 38 kg
  • 36 kg
  • 33 kg
Consumbales
Frequently Bought Together