
2-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:552-89-6
:
Formula:C₇H₅NO₃
MDL Number:MFCD00007132
:151.12 g/mol
Merck Index:12,06684
Boiling Pt:285 °C (1013 hPa)
Melting Pt:41…43 °C
Density:1.35 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
EINECS:209-025-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:41 - 43 °C
- Molar Mass:151.12 g/mol
- Bulk Density:350 kg/m3
- Boiling Point:156 °C (20 hPa)
- Ignition Point:200 °C (Lit.)
Specification Test Results
Related Information
Frequently Bought Together