Order Entry
Poland
ContactUsLinkComponent
 
4-(Dimethylamino)benzaldehyde Reag. Ph. Eur. analytical reagent, Supelco®
 :  Merck
 
undefined
4-(Dimethylamino)benzaldehyde Reag. Ph. Eur. analytical reagent, Supelco®
 :  Merck
 

 

  • Solubility:
    0.3 g/l (20 °C)
  • Melting Point:
    72 - 75 °C
  • Molar Mass:
    149.19 g/mol
  • Bulk Density:
    350 kg/m3
  • Boiling Point:
    176 - 177 °C (23 hPa)
  • Flash Point:
    100 °C
  • Ignition Point:
    445 °C

 

Frequently Bought Together