Order Entry
Poland
Orders LinkContactUsLinkComponent
Water purification system, Milli-Q® Integral
Supplier:   Merck
Water purification system, Milli-Q® Integral
Supplier:  Merck
Certificates
Water purification system, Milli-Q® Integral
6 Options Available Below
Product Details & Documents
The Milli-Q® Integral system is a compact solution that combines the production of Type II (pure) water and Type I (ultrapure) water in a single unit, eliminating the need for a pretreatment stage upstream of the ultrapure water system.
  • Integrated Elix® technology provides high, consistent water quality at optimum operating cost
  • Resistivity and TOC monitoring allows control over both ionic and organic contaminants that can impact results
  • Placed at the outlet of each dispenser, the system's POD Paks have been designed to remove specific types of contaminants
Specifications for Product Family
Specifications
  • Catalog No:
  • 171-0665
  • 171-0739
  • 171-0642
  • 171-0724
  • 171-0733
  • 171-0742
  • 171-0721
  • 171-0661
  • 171-0730
  • 171-0662
  • 171-0769
  • 171-0741
  • 171-0715
  • 171-0732
  • 171-0723
  • 171-0736
  • 171-0727
  • 171-0718
  • 171-0745
  • 171-0668
  • 171-0666
  • 171-0667
  • 171-0726
  • 171-0735
  • 171-0744
  • 171-0716
  • 171-0725
  • 171-0734
  • 171-0743
  • Model:
  • Milli-Q® Integral 3
  • Milli-Q® Integral 10
  • Milli-Q® Integral 3
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 15
  • Milli-Q® Integral 3
  • Milli-Q® Integral 5
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 15
  • Milli-Q® Integral 15
  • Milli-Q® Integral 3
  • Milli-Q® Integral 10
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 5
  • Milli-Q® Integral 3
  • Milli-Q® Integral 15
  • Milli-Q® Integral 15
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 15
  • Milli-Q® Integral 3
  • Milli-Q® Integral 5
  • Milli-Q® Integral 10
  • Milli-Q® Integral 15
  • Flow rate:
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Up to 2,0 l/min (at the E-POD® level)
  • Production flow rate:
  • 3 L/h
  • 10 L/h
  • 3 L/h
  • 5 L/h
  • 10 L/h
  • 15 L/h
  • 3 L/h
  • 5 L/h
  • 5 L/h
  • 10 L/h
  • 15 L/h
  • 15 L/h
  • 3 L/h
  • 10 L/h
  • 5 L/h
  • 10 L/h
  • 5 L/h
  • 3 L/h
  • 15 L/h
  • 15 L/h
  • 5 L/h
  • 10 L/h
  • 5 L/h
  • 10 L/h
  • 15 L/h
  • 3 L/h
  • 5 L/h
  • 10 L/h
  • 15 L/h
  • TOC content:
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • <5 ppb
  • Resistivity at +25 °C:
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • 18,2 MΩ.cm
  • Bacteria:
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • <0,1
  • Particles:
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • <1
  • Pyrogen:
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak® filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak® filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak® filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak® filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • < 0,001 (with BioPak filter as POD Pak)
  • RNases:
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • <0,01
  • DNases:
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • <4
  • W×D×H:
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • 332×484×500 mm
  • Operating weight with full tank:
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
  • 24…28 kg
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 171-0665
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Spanish)
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Spanish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0665
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0739
    Water purification system, Milli-Q® Integral 10 (UK-plug, manual in English)
    Water purification system, Milli-Q® Integral 10 (UK-plug, manual in English)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0739
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak® filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0642
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in French)
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in French)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0642
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0724
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in French)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in French)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0724
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0733
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in French)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in French)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0733
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0742
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in French)
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in French)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0742
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0721
    Water purification system, Milli-Q® Integral 3 (UK-plug, manual in English)
    Water purification system, Milli-Q® Integral 3 (UK-plug, manual in English)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0721
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak® filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0661
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in English)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in English)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 96 570 zł
    96 570 zł
    1 * 1 SZT
       96 570 zł
    Specifications:
    More
    Specifications
    Cat. No.
    171-0661
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0730
    Water purification system, Milli-Q® Integral 5 (UK-plug, manual in English)
    Water purification system, Milli-Q® Integral 5 (UK-plug, manual in English)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0730
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak® filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0662
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in English)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in English)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 113 640 zł
    113 640 zł
    1 * 1 SZT
       113 640 zł
    Specifications:
    More
    Specifications
    Cat. No.
    171-0662
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0769
    Water purification system, Milli-Q® Integral 15 (UK-plug, manual in English)
    Water purification system, Milli-Q® Integral 15 (UK-plug, manual in English)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0769
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak® filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0741
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Danish)
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Danish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0741
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0715
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Danish)
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Danish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0715
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0732
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Danish)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Danish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0732
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0723
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Danish)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Danish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0723
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0736
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Italian)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Italian)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0736
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0727
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Italian)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Italian)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0727
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0718
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Italian)
    Water purification system, Milli-Q® Integral 3 (EU-plug, manual in Italian)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0718
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0745
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Italian)
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Italian)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0745
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0668
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Spanish)
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in Spanish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0668
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0666
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Spanish)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in Spanish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0666
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0667
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Spanish)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in Spanish)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0667
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0726
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in German)
    Water purification system, Milli-Q® Integral 5 (EU-plug, manual in German)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0726
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0735
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in German)
    Water purification system, Milli-Q® Integral 10 (EU-plug, manual in German)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0735
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0744
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in German)
    Water purification system, Milli-Q® Integral 15 (EU-plug, manual in German)
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    171-0744
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0716
    Water purification system, Milli-Q® Integral 3
    Water purification system, Milli-Q® Integral 3
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Specifications:
    More
    Specifications
    Cat. No.
    171-0716
    Model
    Milli-Q® Integral 3
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    3 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0725
    Water purification system, Milli-Q® Integral 5
    Water purification system, Milli-Q® Integral 5
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Specifications:
    More
    Specifications
    Cat. No.
    171-0725
    Model
    Milli-Q® Integral 5
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    5 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0734
    Water purification system, Milli-Q® Integral 5
    Water purification system, Milli-Q® Integral 5
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Specifications:
    More
    Specifications
    Cat. No.
    171-0734
    Model
    Milli-Q® Integral 10
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    10 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
  • 171-0743
    Water purification system, Milli-Q® Integral 15
    Water purification system, Milli-Q® Integral 15
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Specifications:
    More
    Specifications
    Cat. No.
    171-0743
    Model
    Milli-Q® Integral 15
    Flow rate
    Up to 2,0 l/min (at the E-POD® level)
    Production flow rate
    15 L/h
    TOC content
    <5 ppb
    Resistivity at +25 °C
    18,2 MΩ.cm
    Bacteria
    <0,1
    Particles
    <1
    Pyrogen
    < 0,001 (with BioPak filter as POD Pak)
    RNases
    <0,01
    DNases
    <4
    W×D×H
    332×484×500 mm
    Operating weight with full tank
    24…28 kg
Cartridges
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Progard cartridge
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 340 zł
    2 340 zł
    1 * 1 SZT
       2 340 zł
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Start-up kit cartridge
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 655 zł
    655 zł
    1 * 1 SZT
       655 zł
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Quantum® TIX polishing cartridge (contains Ionex resin for trace levels of ions)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 290 zł
    2 290 zł
    1 * 1 SZT
       2 290 zł
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Quantum® TEX polishing cartridge (contains Organex & Ionex resins for trace levels of organics and ions)
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 290 zł
    2 290 zł
    1 * 1 SZT
       2 290 zł
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Vent filter
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 796 zł
    796 zł
    1 * 1 SZT
       796 zł
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • 30 l PE pure water storage reservoir
    30 l PE pure water storage reservoir
    171-0053
    Log in for availability
    1 * 1 SZT - 5 151,11 zł
Product Information
  • Catalog #
    Availability
    Price
  • 60 l PE pure water storage reservoir
    60 l PE pure water storage reservoir
    171-0713
    Log in for availability
    1 * 1 SZT - 7 621,11 zł
Product Information
  • Catalog #
    Availability
    Price
  • ROProtect C
    ROProtect C
    171-0755
    Log in for availability
    1 * 48 Tablet - 430 zł
Product Information
  • Catalog #
    Availability
    Price
  • Bactericidal UV lamp, 254 nm, for Elix / RiOs-DI™ systems
    Bactericidal UV lamp, 254 nm, for Elix / RiOs-DI™ systems
    171-0137
    Log in for availability
    1 * 1 SZT - 1 450 zł
Product Information
  • Catalog #
    Availability
    Price
  • Replacement UV lamp 185/254 nm
    Replacement UV lamp 185/254 nm
    171-0097
    Log in for availability
    1 * 1 SZT - 3 040 zł
Product Information
  • Catalog #
    Availability
    Price
  • System wall-mounting bracket for ultrapure water systems
    System wall-mounting bracket for ultrapure water systems
    171-0133
    Log in for availability
    1 * 1 SZT - 839,17 zł
Product Information
  • Catalog #
    Availability
    Price
  • Wall-mounting bracket for 30, 60 l tank
    Wall-mounting bracket for 30, 60 l tank
    171-0054
    Log in for availability
    1 * 1 SZT - 1 037,61 zł
Product Information
  • Catalog #
    Availability
    Price
  • ASM (Automatic Sanitisation Module): Decontamination module for PE reservoirs with regular UV light emission inside the reservoir to prevent bacteria development
    ASM (Automatic Sanitisation Module): Decontamination module for PE reservoirs with regular UV light emission inside the reservoir to prevent bacteria development
    171-0395
    Log in for availability
    1 * 1 SZT - 4 538,89 zł
Product Information
  • Catalog #
    Availability
    Price
  • Millipak® Express 40 filter unit, 0,22 µm membrane, non sterile, for particulate and bacteria removal
    Millipak® Express 40 filter unit, 0,22 µm membrane, non sterile, for particulate and bacteria removal
    172-0009
    Log in for availability
    1 * 1 SZT - 974 zł
Product Information
  • Catalog #
    Availability
    Price
  • Wall-mounting bracket for 100 l tank
    Wall-mounting bracket for 100 l tank
    171-0890
    Log in for availability
    1 * 1 SZT - 3 683,89 zł
Product Information
  • Catalog #
    Availability
    Price
  • Q-POD® and E-POD® wall-mounting bracket
    Q-POD® and E-POD® wall-mounting bracket
    171-0231
    Log in for availability
    1 * 1 SZT - 2 016,11 zł
Product Information
  • Catalog #
    Availability
    Price
  • 100 L PE pure water storage reservoir
    100 L PE pure water storage reservoir
    171-0714
    Log in for availability
    1 * 1 SZT - 9 869,44 zł