Benzaldehyde ≥99%
Informacje o tym produkcie
Purity: 99+% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299
- Numer CAS:100-52-7
- Wzór:C₆H₅CHO
- Numer MDL:MFCD00003299
- Molecular Weight:106.12 g/mol
- Indeks Mercka:14,1058
- Temperatura wrzenia:178…179 °C
- Temperatura topnienia:–26 °C
- Gęstość:1.047
- Punkt zapalny:62 °C (143 °F)
- Temperatura przechowywania:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
3 dostępnych opcji poniżej
Product Details & Documents
Purity: 99+% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299
- Numer CAS:100-52-7
- Wzór:C₆H₅CHO
- Numer MDL:MFCD00003299
- Molecular Weight:106.12 g/mol
- Indeks Mercka:14,1058
- Temperatura wrzenia:178…179 °C
- Temperatura topnienia:–26 °C
- Gęstość:1.047
- Punkt zapalny:62 °C (143 °F)
- Temperatura przechowywania:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
Documents
Product Family Options
Informacje o produkcie
- Pack typePkDostępnośćPrice
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



